Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010219.3 | NP_034349.1 | 10 | 1445 | ||
GenBank mRNA | AK035395.1 | - | 2 | 135 | ||
AK051433.1 | - | 1564 | ||||
AK083912.1 | - | 10 | 1445 | |||
AK089972.1 | - | 10 | 1454 | |||
AK159211.1 | - | 10 | 1387 | |||
AK160202.1 | - | 10 | 1394 | |||
BC003447.1 | - | 10 | 1462 | 2651490 | ||
X17069.2 | - | 10 | 1311 | |||
X70887.1 | - | 10 | 1342 |