Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010220.4 | NP_034350.1 | 9 | 1214 | ||
GenBank mRNA | AK169795.1 | - | 9 | 1155 | ||
BC015260.1 | - | 9 | 1153 | 4237766 | ||
CF949740.1 | - | 2 | 243 | |||
CT010372.1 | - | 8 | 995 | |||
U16959.1 | - | 9 | 1214 | |||
U36220.1 | - | 9 | 1142 |