Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010222.2 | NP_034352.1 | 3 | 519 | ||
GenBank mRNA | AF040252.1 | - | 3 | 491 | ||
AK138924.1 | - | 3 | 435 | |||
BC032961.1 | - | 3 | 476 | 1347695 |