Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008021.4 | NP_032047.4 | 8 | 1766 | ||
GenBank mRNA | AK049177.1 | - | 8 | 1707 | ||
BC006788.1 | - | 1 | 210 | |||
BC065067.1 | - | 8 | 1766 | 6417437 | ||
BC075723.1 | - | 8 | 1785 | 6515008 | ||
Y11245.1 | - | 8 | 1674 |