Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_180974.4 | NP_851305.2 | 3 | 758 | ||
GenBank mRNA | AK030735.1 | - | 3 | 713 | ||
AK031639.1 | - | 4 | 781 | |||
BC138824.1 | - | 3 | 645 | |||
BC138825.1 | - | 3 | 645 | |||
CB524750.1 | - | 2 | 152 |