Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010225.2 | NP_034355.2 | 2 | 1495 | ||
GenBank mRNA | AK132385.1 | - | 2 | 1496 | ||
BC137947.1 | - | 2 | 1407 | 8860966 | ||
BC137949.1 | - | 2 | 1407 | 8860968 | ||
Y11148.1 | - | 2 | 1202 |