Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130412.1 | NP_001123884.1 | 6 | 830 | ||
NM_015763.4 | NP_056578.2 | 5 | 686 | |||
NM_172950.3 | NP_766538.2 | 5 | 686 | |||
GenBank mRNA | AF180471.1 | - | 5 | 665 | ||
AF412811.1 | - | 4 | 624 | |||
AK014526.1 | - | 4 | 647 | |||
AK019539.1 | - | 5 | 824 | |||
AK030921.1 | - | 5 | 689 | |||
AK141489.1 | - | 5 | 651 | |||
AK154837.1 | - | 6 | 827 | |||
BC042462.1 | - | 4 | 640 | 4211202 |