Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010228.3 | NP_034358.2 | 9 | 1476 | ||
GenBank mRNA | AK005502.1 | - | 9 | 1499 | ||
AK146906.1 | - | 9 | 1501 | |||
AK167497.1 | - | 9 | 1283 | |||
AK167596.1 | - | 9 | 1500 | |||
BC029674.1 | - | 9 | 1300 | 5368921 | ||
D88689.1 | - | 9 | 1473 | |||
D88690.1 | - | 9 | 1473 | |||
L07297.1 | - | 9 | 1248 | |||
X78568.1 | - | 9 | 1464 |