Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-DRB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021983.4 | NP_068818.4 | 2 | 196 | ||
GenBank mRNA | AF361548.1 | - | 2 | 134 | ||
AF361549.1 | - | 2 | 134 | |||
AK292151.1 | - | 2 | 176 | |||
BC005312.1 | - | 2 | 188 | 3933019 | ||
DQ284431.1 | - | 2 | 186 | |||
DQ284433.1 | - | 2 | 187 | |||
GQ891550.1 | - | 2 | 125 | |||
M14190.1 | - | 1 | 38 | |||
M15071.1 | - | 1 | 23 | |||
M15178.1 | - | 2 | 67 | |||
M16732.1 | - | 1 | 25 | |||
M16942.1 | - | 2 | 153 | |||
M17385.1 | - | 1 | 38 | |||
M17388.1 | - | 1 | 38 | |||
U70543.1 | - | 2 | 66 | |||
U70544.1 | - | 2 | 774 |
Gene Symbol: HLA-DRB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001298.1 | 2 | 212 | |||
GenBank mRNA | AF005487.1 | - | 2 | 212 | ||
L76565.1 | - | 2 | 212 | |||
U33547.1 | - | 2 | 232 |