Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276408.1 | NP_001263337.1 | 40 | 6757 | ||
NM_001276409.1 | NP_001263338.1 | 40 | 6682 | |||
NM_001276410.1 | NP_001263339.1 | 39 | 6487 | |||
NM_001276411.1 | NP_001263340.1 | 39 | 6412 | |||
NM_001276412.1 | NP_001263341.1 | 40 | 6397 | |||
NM_001276413.1 | NP_001263342.1 | 39 | 6127 | |||
NM_010233.2 | NP_034363.1 | 41 | 7030 | |||
GenBank mRNA | AK090130.1 | - | 41 | 6982 | ||
AK090135.1 | - | 41 | 6970 | |||
AK133868.1 | - | 15 | 2512 | |||
AK147249.1 | - | 31 | 5107 | |||
AK147315.1 | - | 40 | 6621 | |||
AK147639.1 | - | 40 | 6336 | |||
AK147663.1 | - | 39 | 6350 | |||
AK147683.1 | - | 40 | 6696 | |||
AK147701.1 | - | 39 | 6065 | |||
AK170755.1 | - | 39 | 6455 | |||
BC025521.1 | - | 11 | 1778 | |||
BC036167.1 | - | 14 | 2209 | |||
BC085240.1 | - | 12 | 2097 | |||
BC138421.1 | - | 39 | 6276 | 8861441 | ||
BC145271.1 | - | 39 | 5991 | 9053825 | ||
CN532664.1 | - | 2 | 111 | |||
CN536551.1 | - | 2 | 541 | |||
X93167.1 | - | 24 | 4085 |