Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008044.2 | NP_032070.1 | 4 | 442 | ||
GenBank mRNA | AK148147.1 | - | 4 | 464 | ||
AK160458.1 | - | 4 | 452 | |||
BC016644.1 | - | 3 | 1155 | |||
BC038010.1 | - | 3 | 1166 | |||
BC058533.1 | - | 4 | 467 | |||
BC125337.1 | - | 4 | 440 | 40129852 | ||
BC132337.1 | - | 4 | 440 | 40130614 | ||
U95736.1 | - | 4 | 442 |
Gene Symbol: Gm5037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF223569.1 | - | 1 | 314 |