Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fth1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010239.2 | NP_034369.1 | 1, 2 | 340 | ||
GenBank mRNA | AK027998.1 | - | 1, 2 | 283 | ||
AK139622.1 | - | 1, 2 | 283 | |||
AK147082.1 | - | 1, 2 | 286 | |||
AK150262.1 | - | 1, 2 | 283 | |||
AK150508.1 | - | 1, 2 | 283 | |||
AK150628.1 | - | 1, 2 | 283 | |||
AK150679.1 | - | 1, 2 | 286 | |||
AK150693.1 | - | 1, 2 | 283 | |||
AK151192.1 | - | 1, 2 | 283 | |||
AK151241.1 | - | 1, 2 | 286 | |||
AK151399.1 | - | 1, 2 | 269 | |||
AK151609.1 | - | 1, 2 | 283 | |||
AK151675.1 | - | 1, 2 | 283 | |||
AK152071.1 | - | 1, 2 | 283 | |||
AK152542.1 | - | 1, 2 | 286 | |||
AK152702.1 | - | 2, 3 | 911 | |||
AK153017.1 | - | 1, 2 | 283 | |||
AK153195.1 | - | 1, 2 | 286 | |||
AK153199.1 | - | 1, 2 | 286 | |||
AK159243.1 | - | 1, 2 | 283 | |||
AK168601.1 | - | 1, 2 | 283 | |||
AK169004.1 | - | 1, 2 | 283 | |||
BC011096.1 | - | 1, 2 | 265 | |||
BC012314.1 | - | 1, 2 | 261 | 3488821 | ||
J03941.1 | - | 1, 2 | 236 | |||
X12812.1 | - | 1, 2 | 182 |