Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008052.3 | NP_032078.2 | 5, 6 | 2036 | ||
GenBank mRNA | AB015422.1 | - | 4, 5 | 1639 | ||
AK088630.1 | - | 5, 6 | 1992 | |||
AK169455.1 | - | 5, 6 | 2037 | |||
AK220514.1 | - | 4, 5 | 1617 | |||
BC053055.1 | - | 5, 6 | 1182 | 5685525 | ||
BC065075.1 | - | 6, 7 | 1818 | |||
U38252.1 | - | 3, 4 | 1221 |