Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122892.1 | NP_001116364.1 | 3, 4 | 693 | ||
NM_001122893.1 | NP_001116365.1 | 4, 5 | 763 | |||
NM_008054.2 | NP_032080.2 | 4, 5 | 763 | |||
GenBank mRNA | AK156584.1 | - | 4, 5 | 765 | ||
AK171646.1 | - | 4, 5 | 765 | |||
AK179168.1 | - | 1, 2 | 166 | |||
AK181277.1 | - | 2, 3 | 312 | |||
BC010554.1 | - | 3, 4 | 376 | |||
BC032149.1 | - | 3, 4 | 408 | 3602546 | ||
BC092217.1 | - | 3, 4 | 689 | 6403711 | ||
M27266.1 | - | 4, 5 | 475 | |||
U70324.1 | - | 4, 5 | 476 |