Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122892.1 | NP_001116364.1 | 11, 12 | 1715 | ||
NM_001122893.1 | NP_001116365.1 | 12, 13 | 1794 | |||
NM_008054.2 | NP_032080.2 | 12, 13 | 1785 | |||
GenBank mRNA | AK156584.1 | - | 12, 13 | 1796 | ||
AK171646.1 | - | 12, 13 | 1787 | |||
BC010554.1 | - | 11, 12 | 1396 | |||
BC032149.1 | - | 11, 12 | 1430 | 3602546 | ||
BC092217.1 | - | 11, 12 | 1711 | 6403711 | ||
M27266.1 | - | 13, 14 | 1497 | |||
U70324.1 | - | 12, 13 | 1498 |