Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc37a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293630.1 | NP_001280559.1 | 3 | 824 | ||
NM_001293631.1 | NP_001280560.1 | 3 | 537 | |||
NM_008063.2 | NP_032089.2 | 2 | 564 | |||
GenBank mRNA | AF080469.1 | - | 2 | 385 | ||
AK003620.1 | - | 2 | 387 | |||
AK147849.1 | - | 3 | 494 | |||
AK155339.1 | - | 3 | 821 | |||
BC075655.1 | - | 2 | 564 | 30542293 |