Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159324.1 | NP_001152796.1 | 19 | 2867 | ||
NM_008064.3 | NP_032090.3 | 19 | 2957 | |||
GenBank mRNA | AK052211.1 | - | 19 | 2938 | ||
AK088481.1 | - | 19 | 2933 | |||
AK139333.1 | - | 19 | 2905 | |||
AK141546.1 | - | 19 | 2940 | |||
AK145631.1 | - | 18 | 2374 | |||
AK146538.1 | - | 19 | 2861 | |||
AK150970.1 | - | 19 | 2915 | |||
AK186000.1 | - | 2 | 118 | |||
AK206628.1 | - | 2 | 98 | |||
AK209790.1 | - | 2 | 145 | |||
BC010210.1 | - | 19 | 2921 | 3969127 | ||
U49351.1 | - | 19 | 2809 |