Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162477.1 | NP_001155949.1 | 4 | 1189 | ||
NM_010248.2 | NP_034378.2 | 4 | 1189 | |||
GenBank mRNA | AB018414.1 | - | 4 | 1114 | ||
AF104244.1 | - | 4 | 1189 | |||
BC103524.1 | - | 4 | 1164 | 40039291 | ||
BC103525.1 | - | 4 | 1164 | 40039280 | ||
BC103526.1 | - | 4 | 1164 | 40039281 | ||
BC103527.1 | - | 4 | 1164 | 40039282 |