Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008065.2 | NP_032091.2 | 3 | 590 | ||
GenBank mRNA | AK139916.1 | - | 3 | 270 | ||
AK145446.1 | - | 3 | 270 | |||
AK145838.1 | - | 3 | 335 | |||
AK148087.1 | - | 3 | 361 | |||
AK150455.1 | - | 3 | 270 | |||
AY282793.1 | - | 3 | 597 | |||
AY282794.1 | - | 3 | 256 | |||
AY282795.1 | - | 3 | 472 | |||
BC052448.1 | - | 3 | 590 | 6408997 | ||
M74515.1 | - | 3 | 597 |