Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabpb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271467.1 | NP_001258396.1 | 7 | 969 | ||
NM_001271468.1 | NP_001258397.1 | 6 | 871 | |||
NM_001271470.1 | NP_001258399.1 | 7 | 1036 | |||
NM_001271492.1 | NP_001258421.1 | 6 | 871 | |||
NM_010249.2 | NP_034379.1 | 6 | 871 | |||
NM_207669.2 | NP_997552.1 | 6 | 871 | |||
GenBank mRNA | AK040722.1 | - | 6 | 872 | ||
AK052828.1 | - | 847 | ||||
AK139710.1 | - | 7 | 1036 | |||
AK140222.1 | - | 7 | 922 | |||
AK140907.1 | - | 6 | 782 | |||
AK146373.1 | - | 6 | 735 | |||
AY282803.1 | - | 3 | 328 | |||
AY282804.1 | - | 4 | 410 | |||
AY282805.1 | - | 3 | 411 | |||
BC013558.1 | - | 6 | 762 | 3592119 | ||
CA552206.1 | - | 3 | 288 | |||
CF730595.1 | - | 6 | 724 | |||
M74516.1 | - | 6 | 766 | |||
M74517.1 | - | 6 | 765 |