Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021958.3 | NP_068777.1 | 3 | 1241 | ||
GenBank mRNA | AB208812.1 | - | 1 | 2734 | ||
AK056326.1 | - | 3 | 1238 | |||
AK127923.1 | - | 3 | 1131 | |||
AK313298.1 | - | 3 | 1128 | |||
BC007294.1 | - | 3 | 1226 | 3349581 | ||
BC033808.1 | - | 3 | 999 | 5184776 | ||
M60721.1 | - | 3 | 1197 |