Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077475.1 | - | 9 | 970 | ||
AK154760.1 | - | 9 | 1132 | |||
AK161339.1 | - | 4 | 422 | |||
BC065140.1 | - | 9 | 1005 | |||
BC086671.1 | - | 9 | 1009 | 30635559 | ||
D38557.1 | - | 9 | 1079 |