Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040189.1 | - | 4 | 1018 | ||
AK077475.1 | - | 4 | 373 | |||
AK154760.1 | - | 4 | 534 | |||
BC065140.1 | - | 4 | 407 | |||
BC086671.1 | - | 4 | 411 | 30635559 | ||
D38557.1 | - | 4 | 481 |