Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMBS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000190.3 | NP_000181.2 | 13 | 1020 | ||
NM_001024382.1 | NP_001019553.1 | 13 | 922 | |||
NM_001258208.1 | NP_001245137.1 | 12 | 900 | |||
NM_001258209.1 | NP_001245138.1 | 12 | 991 | |||
XM_005271531.1 | XP_005271588.1 | 13 | 1098 | |||
XM_005271532.1 | XP_005271589.1 | 13 | 1076 | |||
XM_005271533.3 | XP_005271590.1 | 12 | 972 | |||
XM_011542796.1 | XP_011541098.1 | 12 | 1352 | |||
XM_017017629.1 | XP_016873118.1 | 13 | 1486 | |||
GenBank mRNA | AK000628.1 | - | 12 | 983 | ||
AK131072.1 | - | 12 | 863 | |||
AK290275.1 | - | 13 | 1016 | |||
AK310343.1 | - | 1360 | ||||
BC000520.2 | - | 13 | 1014 | 2822949 | ||
BC008149.1 | - | 13 | 975 | 4178944 | ||
BC019323.1 | - | 13 | 1014 | 2822949 | ||
BX647328.1 | - | 10 | 1625 | |||
X04217.1 | - | 13 | 893 | |||
X04808.1 | - | 13 | 890 |