Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015736.2 | NP_056551.2 | 3 | 943 | ||
GenBank mRNA | AK135489.1 | - | 3 | 904 | ||
BC043331.1 | - | 3 | 799 | 5342768 | ||
U70538.1 | - | 3 | 787 |