Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK088379.1 | - | 7 | 693 | ||
AK134525.1 | - | 4 | 348 | |||
AV027352.2 | - | 1 | 131 | |||
BC010985.1 | - | 7 | 698 | 4038194 | ||
CX239267.1 | - | 7 | 691 | |||
M96265.1 | - | 7 | 725 |