Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109657.3 | NP_001103127.1 | 11 | 1213 | ||
NM_001277079.1 | NP_001264008.1 | 11 | 1042 | |||
NM_001277080.1 | NP_001264009.1 | 10 | 956 | |||
NM_008088.5 | NP_032114.2 | 11 | 1019 | |||
GenBank mRNA | AF133184.1 | - | 7 | 891 | ||
AK017394.1 | - | 11 | 1012 | |||
AK076926.1 | - | 12 | 1095 | |||
AK152149.1 | - | 9 | 716 | |||
AK154463.1 | - | 11 | 1016 | |||
AK172943.1 | - | 7 | 835 | |||
BC120528.1 | - | 11 | 940 | 40129451 | ||
BC120530.1 | - | 11 | 940 | 40129453 | ||
CV562268.1 | - | 3 | 257 | |||
U19860.1 | - | 11 | 1045 |