Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008092.3 | NP_032118.2 | 4 | 1472 | ||
GenBank mRNA | AB075549.1 | - | 4 | 1490 | ||
AF179424.1 | - | 3 | 893 | |||
AK134639.1 | - | 3 | 496 | |||
BC137824.1 | - | 3 | 1225 | 8860843 | ||
M98339.1 | - | 4 | 1440 | |||
U85046.1 | - | 4 | 1443 |