Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301628.1 | NP_001288557.1 | 11, 12 | 1574 | ||
NM_027604.4 | NP_081880.2 | 12, 13 | 1661 | |||
GenBank mRNA | AF468037.1 | - | 12, 13 | 1565 | ||
AK014891.1 | - | 6, 7 | 872 | |||
AK122301.1 | - | 11, 12 | 1489 | |||
AK133852.1 | - | 11, 12 | 1483 | |||
AK145749.1 | - | 12, 13 | 1565 | |||
AK161469.1 | - | 12, 13 | 1604 | |||
AK161957.1 | - | 11, 12 | 1522 | |||
AK163315.1 | - | 11, 12 | 1503 | |||
AK168755.1 | - | 11, 12 | 1509 | |||
BC050042.1 | - | 12, 13 | 1577 | 30022307 |