Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301628.1 | NP_001288557.1 | 14, 15 | 1983 | ||
NM_027604.4 | NP_081880.2 | 15, 16 | 2070 | |||
GenBank mRNA | AF468037.1 | - | 15, 16 | 1974 | ||
AK014891.1 | - | 9, 10 | 1281 | |||
AK122301.1 | - | 14, 15 | 1898 | |||
AK133852.1 | - | 14, 15 | 1892 | |||
AK145749.1 | - | 15, 16 | 1974 | |||
AK161469.1 | - | 15, 16 | 2013 | |||
AK161957.1 | - | 14, 15 | 1931 | |||
AK163315.1 | - | 14, 15 | 1912 | |||
AK168755.1 | - | 14, 15 | 1918 | |||
AK216706.1 | - | 1, 2 | 73 | |||
BC003978.1 | - | 2, 3 | 329 | |||
BC050042.1 | - | 15, 16 | 1986 | 30022307 |