Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301228.1 | NP_001288157.1 | 2 | 316 | ||
NM_001301229.1 | NP_001288158.1 | 2 | 207 | |||
NM_001301231.1 | NP_001288160.1 | 2 | 246 | |||
NM_005342.3 | NP_005333.2 | 2 | 211 | |||
GenBank mRNA | BC010378.1 | - | 1 | 131 | ||
BC037210.1 | - | 1 | 116 | |||
BC070482.1 | - | 3 | 295 | 6140626 | ||
BF979310.1 | - | 2 | 252 | |||
BX537505.1 | - | 2 | 196 | |||
DB191695.1 | - | 2 | 196 | |||
DN996967.1 | - | 2 | 218 | |||
L05085.1 | - | 1 | 116 | |||
Y10043.1 | - | 2 | 190 |
Gene Symbol: HMGB3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002165.1 | 1 | 101 | |||
GenBank mRNA | BC103849.1 | - | 1 | 101 | ||
BC103850.1 | - | 1 | 101 | |||
BC103851.1 | - | 1 | 101 |