Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038010.2 | NP_001033099.1 | 5 | 905 | ||
NM_020004.5 | NP_064388.2 | 5 | 905 | |||
GenBank mRNA | AF254441.1 | - | 5 | 842 | ||
AK137358.1 | - | 5 | 882 | |||
AK145205.1 | - | 4 | 803 | |||
AK147042.1 | - | 5 | 853 | |||
AK150225.1 | - | 5 | 883 | |||
AK154866.1 | - | 5 | 893 | |||
AK155347.1 | - | 5 | 886 | |||
AK155791.1 | - | 5 | 906 | |||
AK155842.1 | - | 5 | 906 | |||
AK158079.1 | - | 5 | 893 | |||
AK202408.1 | - | 2 | 242 | |||
BC011088.1 | - | 5 | 611 | |||
BC051952.1 | - | 5 | 887 | |||
BC060050.1 | - | 5 | 887 | |||
BC063752.1 | - | 5 | 890 | 6333090 |