Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004965.6 | NP_004956.5 | 4 | 304 | ||
XM_011529543.1 | XP_011527845.1 | 4 | 282 | |||
XM_011529544.1 | XP_011527846.1 | 4 | 282 | |||
XM_011529545.1 | XP_011527847.1 | 3 | 85 | |||
XM_011529547.1 | XP_011527849.1 | 3 | 85 | |||
XM_011529549.1 | XP_011527851.1 | 4 | 297 | |||
XM_017028332.1 | XP_016883821.1 | 4 | 247 | |||
XM_017028333.1 | XP_016883822.1 | 4 | 282 | |||
XM_017028334.1 | XP_016883823.1 | 4 | 234 | |||
XM_017028335.1 | XP_016883824.1 | 3 | 85 | |||
XM_017028336.1 | XP_016883825.1 | 4 | 282 | |||
XM_017028338.1 | XP_016883827.1 | 4 | 297 | |||
GenBank mRNA | AB209245.1 | - | 4 | 237 | ||
AK315410.1 | - | 4 | 247 | |||
BC000075.2 | - | 4 | 224 | 3510312 | ||
BC023984.1 | - | 4 | 216 | 4104558 | ||
BC070153.1 | - | 4 | 242 | 6450636 | ||
BC070154.1 | - | 4 | 222 | 6495327 | ||
BC071643.1 | - | 4 | 212 | 5549431 | ||
BC072672.1 | - | 4 | 213 | 6166317 | ||
BC106080.1 | - | 4 | 237 | 3855939 | ||
BE779939.1 | - | 363 | ||||
BT007337.1 | - | 1 | 100 | |||
J02621.1 | - | 4 | 250 |
Gene Symbol: ACTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX641076.1 | - | 1 | 280 |
Gene Symbol: NBR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003108.1 | 5 | 1313 | |||
GenBank mRNA | CR936675.1 | - | 5 | 1184 |