Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136484.3 | NP_001129956.3 | 3 | 1212 | ||
NM_010265.5 | NP_034395.4 | 3 | 1253 | |||
NM_173442.5 | NP_775618.3 | 3 | 1285 | |||
GenBank mRNA | AK045216.1 | - | 3 | 1236 | ||
BC039126.1 | - | 3 | 1210 | 3154671 | ||
D87332.1 | - | 3 | 1254 | |||
D87333.1 | - | 3 | 1236 | |||
U19265.1 | - | 4 | 1480 |