Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010269.3 | NP_034399.1 | 13 | 1625 | ||
GenBank mRNA | AK004887.1 | - | 13 | 1571 | ||
AK082958.1 | - | 13 | 1587 | |||
AK146835.1 | - | 13 | 1615 | |||
AK161122.1 | - | 13 | 1524 | |||
AK166799.1 | - | 13 | 1540 | |||
AK166859.1 | - | 13 | 1539 | |||
BC010192.1 | - | 2 | 78 | |||
BC025070.1 | - | 13 | 1534 | 3710712 | ||
Y17851.1 | - | 13 | 1621 |