Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008112.4 | NP_032138.3 | 5 | 654 | ||
GenBank mRNA | AK007871.1 | - | 5 | 597 | ||
AK013104.1 | - | 5 | 599 | |||
AK078144.1 | - | 597 | ||||
AK078514.1 | - | 5 | 601 | |||
AK079690.1 | - | 4 | 495 | |||
AK083063.1 | - | 5 | 596 | |||
AK090083.1 | - | 5 | 602 | |||
AK137761.1 | - | 5 | 601 | |||
AK143633.1 | - | 5 | 599 | |||
AK143681.1 | - | 5 | 633 | |||
AK149958.1 | - | 5 | 607 | |||
AK150356.1 | - | 5 | 589 | |||
AK150655.1 | - | 5 | 624 | |||
AK152250.1 | - | 5 | 588 | |||
AK153381.1 | - | 5 | 567 | |||
AK159734.1 | - | 5 | 599 | |||
AK166317.1 | - | 5 | 599 | |||
AK167659.1 | - | 5 | 611 | |||
AK169299.1 | - | 5 | 565 | |||
AK207166.1 | - | 2 | 144 | |||
AK213780.1 | - | 2 | 143 | |||
BC024971.1 | - | 5 | 575 | |||
BC053381.1 | - | 2754 | ||||
BC055341.1 | - | 5 | 580 | 6808475 | ||
L36314.1 | - | 5 | 460 |