Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgdig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008113.3 | NP_032139.1 | 4 | 574 | ||
GenBank mRNA | AK144321.1 | - | 4 | 426 | ||
BC002032.1 | - | 4 | 571 | 3490167 | ||
BC158003.1 | - | 4 | 439 | 9007312 | ||
L42463.1 | - | 4 | 405 | |||
U73198.1 | - | 4 | 414 |