Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145820.1 | NP_001139292.1 | 16 | 2334 | ||
NM_010274.3 | NP_034404.3 | 16 | 2209 | |||
GenBank mRNA | AB045714.1 | - | 3 | 187 | ||
AK034353.1 | - | 16 | 2118 | |||
AK144716.1 | - | 16 | 2335 | |||
AK151189.1 | - | 16 | 2122 | |||
AK167152.1 | - | 16 | 2147 | |||
BC021359.1 | - | 16 | 2303 | 5012142 | ||
D50430.1 | - | 16 | 2111 | |||
U60987.1 | - | 16 | 2145 |