Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013529.3 | NP_038557.1 | 11 | 1121 | ||
GenBank mRNA | AB016780.1 | - | 11 | 1087 | ||
AK132821.1 | - | 10 | 967 | |||
BC031928.1 | - | 11 | 1089 | 4167189 |