Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013529.3 | NP_038557.1 | 15 | 1576 | ||
GenBank mRNA | AB016780.1 | - | 15 | 1542 | ||
AK132821.1 | - | 14 | 1422 | |||
BC031928.1 | - | 15 | 1544 | 4167189 |