Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013529.3 | NP_038557.1 | 5 | 452 | ||
GenBank mRNA | AB016780.1 | - | 5 | 418 | ||
AK132821.1 | - | 4 | 298 | |||
BC031928.1 | - | 5 | 420 | 4167189 |