Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285457.1 | NP_001272386.1 | 5 | 1063 | ||
NM_010279.3 | NP_034409.1 | 7 | 1279 | |||
GenBank mRNA | AB000800.1 | - | 6 | 833 | ||
AF012811.1 | - | 2 | 105 | |||
AF015172.1 | - | 5 | 1046 | |||
AK076171.1 | - | 7 | 1200 | |||
AK139987.1 | - | 2 | 119 | |||
BC011532.1 | - | 5 | 1010 | |||
BC040251.1 | - | 5 | 847 | |||
BC044783.2 | - | 7 | 1159 | |||
BC054378.1 | - | 6 | 950 | 6390018 | ||
KM079499.1 | - | 6 | 938 |