Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010281.2 | NP_034411.2 | 2 | 299 | ||
GenBank mRNA | AF051102.1 | - | 2 | 267 | ||
AK089996.1 | - | 2 | 256 | |||
BB610405.1 | - | 2 | 187 | |||
BC009809.1 | - | 2 | 190 | 4223953 | ||
BC150898.1 | - | 2 | 236 | 9087809 | ||
BE859353.1 | - | 2 | 192 |