Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF297615.1 | - | 6, 7 | 692 | ||
AK004753.1 | - | 6, 7 | 755 | |||
AK159087.1 | - | 6, 7 | 764 | |||
M26925.1 | - | 5, 6 | 536 | |||
M85153.1 | - | 6, 7 | 635 |