Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022305.4 | NP_071641.1 | 6 | 1816 | ||
GenBank mRNA | AK131721.1 | - | 6 | 919 | ||
AK132970.1 | - | 6 | 1817 | |||
AK154393.1 | - | 6 | 1275 | |||
AK167322.1 | - | 6 | 1075 | |||
BC053006.1 | - | 6 | 1277 | 5698356 | ||
D00314.1 | - | 6 | 1230 | |||
J03880.1 | - | 6 | 1255 |