Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022305.4 | NP_071641.1 | 3 | 1402 | ||
GenBank mRNA | AK029474.1 | - | 3 | 655 | ||
AK131721.1 | - | 3 | 505 | |||
AK132970.1 | - | 3 | 1403 | |||
AK154393.1 | - | 3 | 861 | |||
AK167322.1 | - | 3 | 661 | |||
AK210987.1 | - | 1 | 32 | |||
AK214993.1 | - | 1 | 32 | |||
BC053006.1 | - | 3 | 863 | 5698356 | ||
D00314.1 | - | 3 | 816 | |||
J03880.1 | - | 3 | 841 |