Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008119.2 | NP_032145.2 | 3 | 291 | ||
GenBank mRNA | AK008308.1 | - | 3 | 291 | ||
AK008525.1 | - | 2 | 147 | |||
AK172404.1 | - | 1 | 68 | |||
BC104314.1 | - | 3 | 281 | 40044479 | ||
BC104315.1 | - | 3 | 281 | 40044480 | ||
U34295.1 | - | 2 | 194 |