Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gja8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008123.3 | NP_032149.1 | 2 | 838 | ||
GenBank mRNA | BC120513.1 | - | 2 | 759 | 40129436 | |
BC125569.1 | - | 2 | 759 | 40130084 | ||
GU363513.1 | - | 2 | 717 | |||
GU363514.1 | - | 2 | 807 | |||
GU363515.1 | - | 1 | 1024 |