Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gja8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008123.3 | NP_032149.1 | 2 | 341 | ||
GenBank mRNA | BB665296.1 | - | 2 | 220 | ||
BC120513.1 | - | 2 | 262 | 40129436 | ||
BC125569.1 | - | 2 | 262 | 40130084 | ||
GU363513.1 | - | 2 | 220 | |||
GU363514.1 | - | 2 | 310 | |||
GU363515.1 | - | 1 | 527 |