Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gjd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010290.2 | NP_034420.2 | 2 | 708 | ||
GenBank mRNA | BC058595.1 | - | 2 | 708 | 5364942 |